Learn More
Cayman Chemical JWH 073 4mthylnphthyl nl 10mg
SDP
Catalog No. 502892823
Encompass_Preferred
Differs structurally from JWH 073 by having a methyl group added at the 4 position of the naphthyl rings; a similar addition to JWH 018 results in an analog which has higher affinities for both cannabinoid receptors
Catalog No. 50-289-2823 Supplier Cayman Chemical Supplier No. 900107610 MG
Differs structurally from JWH 073 by having a methyl group added at the 4 position of the naphthyl rings; a similar addition to JWH 018 results in an analog which has higher affinities for both cannabinoid receptors