Learn More
Cayman Chemical JWH 073 6-hydroxyindole metabolite, Purity: ≥95%, CAS: 1307803-48-0, 10 mg
SDP
Catalog No. 502894615
Encompass_Preferred
Expected urinary metabolite of JWH 073
Catalog No. 50-289-4615 Supplier Cayman Chemical Supplier No. 900086310 MG
Expected urinary metabolite of JWH 073