Learn More
Cayman Chemical (±)-JWH 073 N-(3-hydroxybutyl) metabolite, Purity: ≥98%, CAS: 1320363-48-1, 5 mg
SDP
Catalog No. 502889297
Encompass_Preferred
Expected product of the metabolism of JWH 073 by human liver microsomes in vitro, generated by the oxidation of the aminoalkyl chain; biological actions are unknown; intended for research and forensic applications
Catalog No. 50-288-9297 Supplier Cayman Chemical Supplier No. 107955 MG
Expected product of the metabolism of JWH 073 by human liver microsomes in vitro, generated by the oxidation of the aminoalkyl chain; biological actions are unknown; intended for research and forensic applications