Learn More
Cayman Chemical JWH 073 N 3hydroxybutyl meta
SDP
Supplier:  Cayman Chemical 107951 MG
Encompass_Preferred
Expected product of the metabolism of JWH 073 by human liver microsomes in vitro, generated by the oxidation of the aminoalkyl chain; biological actions are unknown; intended for research and forensic applications
Catalog No. 50-288-9343