Learn More
Cayman Chemical JWH 073 N-(4-hydroxybutyl) metabolite, Purity: ≥98%, CAS: 335161-14-3, 10 mg
SDP
Catalog No. 502894516
Encompass_Preferred
Expected to be a major urinary metabolite of JWH 073; intended for research and forensic applications
Catalog No. 50-289-4516 Supplier Cayman Chemical Supplier No. 900086510 MG
Expected to be a major urinary metabolite of JWH 073; intended for research and forensic applications