Learn More
Cayman Chemical JWH 073 N 4hydroxybutyl me
SDP
Supplier:  Cayman Chemical 10934500 UG
Encompass_Preferred
An internal standard for the quantification of JWH 073 N-(4-hydroxybutyl) metabolite by GC- or LC-MS
Catalog No. 50-295-1378