Learn More
Cayman Chemical JWH 073 N 4hydroxybutyl me
SDP
Catalog No. 502951637
Encompass_Preferred
An internal standard for the quantification of JWH 073 N-(4-hydroxybutyl) metabolite by GC- or LC-MS
Catalog No. 50-295-1637 Supplier Cayman Chemical Supplier No. 10934100 UG
An internal standard for the quantification of JWH 073 N-(4-hydroxybutyl) metabolite by GC- or LC-MS