Learn More
Cayman Chemical JWH 073 NbutnoIc ACD metab
SDP
Supplier:  Cayman Chemical 9000870500 UG
Encompass_Preferred
An internal standard for the quanitification of JWH 073 N-butanoic acid metabolite by GC- or LC-MS; intended for research and forensic applications
Catalog No. 50-289-3690