Learn More
Cayman Chemical JWH 081 2methoxynaphthyl5mg
SDP
Catalog No. 502896096
Encompass_Preferred
Differs structurally from JWH 081 by having the methoxy group attached to the naphthyl rings at the 2' position
Catalog No. 50-289-6096 Supplier Cayman Chemical Supplier No. 90010445 MG
Differs structurally from JWH 081 by having the methoxy group attached to the naphthyl rings at the 2' position