Learn More
Cayman Chemical JWH 081 3methoxynaphthyl10mg
SDP
Catalog No. 502895729
Encompass_Preferred
Differs structurally from JWH 081 by having the methoxy group attached to the naphthyl rings at the 3 position, instead of the 4 position; intended for forensic and research purposes
Catalog No. 50-289-5729 Supplier Cayman Chemical Supplier No. 900104510 MG
Differs structurally from JWH 081 by having the methoxy group attached to the naphthyl rings at the 3 position, instead of the 4 position; intended for forensic and research purposes