Learn More
Cayman Chemical JWH 081 3-methoxynaphthyl isomer, Purity: ≥98%, CAS: 1696409-36-5, 1 mg
SDP
Catalog No. 502894299
Encompass_Preferred
Differs structurally from JWH 081 by having the methoxy group attached to the naphthyl rings at the 3 position, instead of the 4 position; intended for forensic and research purposes
Catalog No. 50-289-4299 Supplier Cayman Chemical Supplier No. 90010451 MG
Differs structurally from JWH 081 by having the methoxy group attached to the naphthyl rings at the 3 position, instead of the 4 position; intended for forensic and research purposes