Learn More
Cayman Chemical JWH 081 4-hydroxynaphthyl metabolite, Purity: ≥98%, CAS: 1393345-76-0, 10 mg
SDP
Catalog No. 502952807
Encompass_Preferred
Expected metabolite of JWH 081 that would be detectable both in serum and in urine; intended for research and forensic purposes
Catalog No. 50-295-2807 Supplier Cayman Chemical Supplier No. 1434710 MG
Expected metabolite of JWH 081 that would be detectable both in serum and in urine; intended for research and forensic purposes