Learn More
Cayman Chemical JWH 081 5methoxynaphthyl10mg
SDP
Catalog No. 502895861
Encompass_Preferred
Differs structurally from JWH 081 by having the methoxy group attached to the naphthyl rings at the 5 position, instead of the 4 position
Catalog No. 50-289-5861 Supplier Cayman Chemical Supplier No. 900104610 MG
Differs structurally from JWH 081 by having the methoxy group attached to the naphthyl rings at the 5 position, instead of the 4 position