Learn More
Cayman Chemical JWH 081 5-methoxynaphthyl isomer, Purity: ≥98%, CAS: 1427325-65-2, 1 mg
SDP
Catalog No. 502894300
Encompass_Preferred
Differs structurally from JWH 081 by having the methoxy group attached to the naphthyl rings at the 5 position, instead of the 4 position
Catalog No. 50-289-4300 Supplier Cayman Chemical Supplier No. 90010461 MG
Differs structurally from JWH 081 by having the methoxy group attached to the naphthyl rings at the 5 position, instead of the 4 position