Learn More
Cayman Chemical JWH 081 5methoxynaphthyl5mg
SDP
Catalog No. 502894784
Encompass_Preferred
Differs structurally from JWH 081 by having the methoxy group attached to the naphthyl rings at the 5 position, instead of the 4 position
Catalog No. 50-289-4784 Supplier Cayman Chemical Supplier No. 90010465 MG
Differs structurally from JWH 081 by having the methoxy group attached to the naphthyl rings at the 5 position, instead of the 4 position