Learn More
Cayman Chemical JWH 081 6-methoxynaphthyl isomer, Purity: ≥98%, CAS: 824961-41-3, 10 mg
SDP
Catalog No. 502895305
Encompass_Preferred
Differs structurally from JWH 081 by having the methoxy group attached to the naphthyl rings at the 6 position, instead of the 4 position; intended for forensic and research purposes
Catalog No. 50-289-5305 Supplier Cayman Chemical Supplier No. 900104710 MG
Differs structurally from JWH 081 by having the methoxy group attached to the naphthyl rings at the 6 position, instead of the 4 position; intended for forensic and research purposes