Learn More
Cayman Chemical JWH 081 7-methoxynaphthyl isomer, Purity: ≥98%, CAS: 824961-61-7, 1 mg
SDP
Catalog No. 502892628
Encompass_Preferred
Differs structurally from JWH 081 by having the methoxy group attached to the naphthyl rings at the 7 position, instead of the 4 position
Catalog No. 50-289-2628 Supplier Cayman Chemical Supplier No. 90010481 MG
Differs structurally from JWH 081 by having the methoxy group attached to the naphthyl rings at the 7 position, instead of the 4 position