Learn More
Cayman Chemical JWH 081 8-methoxynaphthyl isomer, Purity: ≥98%, CAS: 1349837-48-4, 5 mg
SDP
Catalog No. 502892630
Encompass_Preferred
Differs structurally from JWH 081 by having the methoxy group attached to the naphthyl rings at the 8 position, instead of the 4 position; intended for forensic and research purposes
Catalog No. 50-289-2630 Supplier Cayman Chemical Supplier No. 90010495 MG
Differs structurally from JWH 081 by having the methoxy group attached to the naphthyl rings at the 8 position, instead of the 4 position; intended for forensic and research purposes