Learn More
Cayman Chemical JWH 081 N-(4-hydroxypentyl) metabolite, Purity: ≥98%, CAS: 2748622-62-8, 10 mg
SDP
Catalog No. 502953912
Encompass_Preferred
An expected product of the phase I metabolism of JWH 081, based on studies on the metabolism of the related synthetic CBs JWH 018 and JWH 073; intended for research and forensic purposes
Catalog No. 50-295-3912 Supplier Cayman Chemical Supplier No. 1434810 MG
An expected product of the phase I metabolism of JWH 081, based on studies on the metabolism of the related synthetic CBs JWH 018 and JWH 073; intended for research and forensic purposes