Learn More
Cayman Chemical JWH 081 N-(5-hydroxypentyl) metabolite, Purity: ≥95%, CAS: 1427325-66-3, 10 mg
SDP
Catalog No. 502895599
Encompass_Preferred
Expected metabolite of JWH 081 that would be detectable both in serum and in urine; intended for research and forensic applications
Catalog No. 50-289-5599 Supplier Cayman Chemical Supplier No. 900076810 MG
Expected metabolite of JWH 081 that would be detectable both in serum and in urine; intended for research and forensic applications