Learn More
Cayman Chemical JWH 081 N-(5-hydroxypentyl) metabolite-d5, Purity: ≥99% deuterated forms (d1-d5), CAS: 2748469-56-7, 1 mg
SDP
Catalog No. 502954118
Encompass_Preferred
An internal standard for the quantification of JWH 081 N-(5-hydroxypentyl) metabolite by GC- or LC-MS; intended for research and forensic applications
Catalog No. 50-295-4118 Supplier Cayman Chemical Supplier No. 143611 MG
An internal standard for the quantification of JWH 081 N-(5-hydroxypentyl) metabolite by GC- or LC-MS; intended for research and forensic applications