Learn More
Cayman Chemical JWH 081 N 5hydroxypentyl m 1mg
SDP
Catalog No. 502895229
Encompass_Preferred
Expected metabolite of JWH 081 that would be detectable both in serum and in urine; intended for research and forensic applications
Catalog No. 50-289-5229 Supplier Cayman Chemical Supplier No. 90007681 MG
Expected metabolite of JWH 081 that would be detectable both in serum and in urine; intended for research and forensic applications