Learn More
Cayman Chemical JWH 081 N 5hydroxypentyl m
SDP
Catalog No. 502953862
Encompass_Preferred
An internal standard for the quantification of JWH 081 N-(5-hydroxypentyl) metabolite by GC- or LC-MS; intended for research and forensic applications
Catalog No. 50-295-3862 Supplier Cayman Chemical Supplier No. 14361500 UG
An internal standard for the quantification of JWH 081 N-(5-hydroxypentyl) metabolite by GC- or LC-MS; intended for research and forensic applications