Learn More
Cayman Chemical JWH 081 N-pentanoic acid metabolite, Purity: ≥98%, CAS: 1537889-08-9, 10 mg
SDP
Catalog No. 502955006
Encompass_Preferred
Expected metabolite of JWH 081 that would be detectable both in serum and in urine
Catalog No. 50-295-5006 Supplier Cayman Chemical Supplier No. 1435310 MG
Expected metabolite of JWH 081 that would be detectable both in serum and in urine