Learn More
Cayman Chemical JWH 081 NpentnoIc ACD meta
SDP
Catalog No. 502953863
Encompass_Preferred
An internal standard for the quantification of JWH 081 N-pentanoic acid metabolite by GC- or LC-MS
Catalog No. 50-295-3863 Supplier Cayman Chemical Supplier No. 14362500 UG
An internal standard for the quantification of JWH 081 N-pentanoic acid metabolite by GC- or LC-MS