Learn More
Cayman Chemical JWH 081 NpentnoIc ACD meta
SDP
Supplier:  Cayman Chemical 14362100 UG
Encompass_Preferred
An internal standard for the quantification of JWH 081 N-pentanoic acid metabolite by GC- or LC-MS
Catalog No. 50-295-5007