Learn More
Cayman Chemical JWH 122 5mthylnaphthyl 10mg
SDP
Catalog No. 502895114
Encompass_Preferred
Differs from JWH 122 in that the methyl group is attached to the naphthyl rings at the 5, rather than the 4, position; intended for forensic purposes
Catalog No. 50-289-5114 Supplier Cayman Chemical Supplier No. 900103410 MG
Differs from JWH 122 in that the methyl group is attached to the naphthyl rings at the 5, rather than the 4, position; intended for forensic purposes