Learn More
Cayman Chemical JWH 122 6-methylnaphthyl isomer, Purity: ≥95%, CAS: 1427325-68-5, 10 mg
SDP
Catalog No. 502892920
Encompass_Preferred
Differs from JWH 122 in that the methyl group is attached to the naphthyl rings at the 6, rather than the 4, position
Catalog No. 50-289-2920 Supplier Cayman Chemical Supplier No. 900103510 MG
Differs from JWH 122 in that the methyl group is attached to the naphthyl rings at the 6, rather than the 4, position