Learn More
Cayman Chemical JWH 122 7-methylnaphthyl isomer, Purity: ≥98%, CAS: 824960-56-7, 10 mg
SDP
Catalog No. 502893694
Encompass_Preferred
Differs from JWH 122 in that the methyl group is attached to the naphthyl rings at the 7, rather than the 4, position
Catalog No. 50-289-3694 Supplier Cayman Chemical Supplier No. 900103610 MG
Differs from JWH 122 in that the methyl group is attached to the naphthyl rings at the 7, rather than the 4, position