Learn More
Cayman Chemical JWH 122 8-methylnaphthyl isomer, Purity: ≥98%, 10 mg
SDP
Catalog No. 502895112
Encompass_Preferred
Differs from JWH 122 in that the methyl group is attached to the naphthyl rings at the 8, rather than the 4, position
Catalog No. 50-289-5112 Supplier Cayman Chemical Supplier No. 900103710 MG
Differs from JWH 122 in that the methyl group is attached to the naphthyl rings at the 8, rather than the 4, position