Learn More
Cayman Chemical JWH 122 N-(5-hydroxypentyl) metabolite, Purity: ≥98%, CAS: 1379604-68-8, 5 mg
SDP
Catalog No. 502951290
Encompass_Preferred
A metabolite of JWH 122 that is characterized by monohydroxylation of the N-alkyl chain
Catalog No. 50-295-1290 Supplier Cayman Chemical Supplier No. 109255 MG
A metabolite of JWH 122 that is characterized by monohydroxylation of the N-alkyl chain