Learn More
Cayman Chemical JWH 133 Analytical Reference S
SDP
Supplier:  Cayman Chemical 1000542825 MG
Encompass_Preferred
An analytical reference standard; a synthetic CB that is selective for the peripheral CB2 receptor (Ki = 3.4 nM) over the central CB1 receptor (Ki = 677 nM); inhibits the growth of tumors in mice, increasing apoptosis and impairing angiogenesis; suppresses inflammation in experimental colitis and pancreatitis in mice
Catalog No. 50-288-8744