Learn More
Cayman Chemical JWH 133 Analytical Reference S
SDP
Catalog No. 502888500
Encompass_Preferred
An analytical reference standard; a synthetic CB that is selective for the peripheral CB2 receptor (Ki = 3.4 nM) over the central CB1 receptor (Ki = 677 nM); inhibits the growth of tumors in mice, increasing apoptosis and impairing angiogenesis; suppresses inflammation in experimental colitis and pancreatitis in mice
Catalog No. 50-288-8500 Supplier Cayman Chemical Supplier No. 100054285 MG
An analytical reference standard; a synthetic CB that is selective for the peripheral CB2 receptor (Ki = 3.4 nM) over the central CB1 receptor (Ki = 677 nM); inhibits the growth of tumors in mice, increasing apoptosis and impairing angiogenesis; suppresses inflammation in experimental colitis and pancreatitis in mice