Learn More
Cayman Chemical JWH 147 Neurochemicals
SDP
Supplier:  Cayman Chemical 1082610 MG
Encompass_Preferred
A synthetic CB with a high affinity for both the central CB1 receptor (Ki = 11 nM) and the peripheral CB2 receptor (Ki = 7.1 nM); intended for research and forensic applications
Catalog No. 50-289-1380