Learn More
Cayman Chemical JWH 203 4-chlorophenyl isomer, Purity: ≥98%, CAS: 864445-58-9, 10 mg
SDP
Catalog No. 502951169
Encompass_Preferred
An analytical reference standard categorized as a synthetic cannabinoid; reduces spontaneous locomotor activity, but does not induce antinociception or a decrease in body temperature, in mice; intended for research and forensic applications
Catalog No. 50-295-1169 Supplier Cayman Chemical Supplier No. 1086710 MG
An analytical reference standard categorized as a synthetic cannabinoid; reduces spontaneous locomotor activity, but does not induce antinociception or a decrease in body temperature, in mice; intended for research and forensic applications