Learn More
Cayman Chemical JWH 203 N 5hydroxypentyl m
SDP
Supplier:  Cayman Chemical 14368100 UG
Encompass_Preferred
An internal standard for the quantification of JWH 203 N-(5-hydroxypentyl) metabolite by GC- or LC-MS
Catalog No. 50-297-0046