Learn More
Cayman Chemical JWH 203 N 5hydroxypentyl m
SDP
Catalog No. 502970660
Encompass_Preferred
An internal standard for the quantification of JWH 203 N-(5-hydroxypentyl) metabolite by GC- or LC-MS
Catalog No. 50-297-0660 Supplier Cayman Chemical Supplier No. 14368500 UG
An internal standard for the quantification of JWH 203 N-(5-hydroxypentyl) metabolite by GC- or LC-MS