Learn More
Cayman Chemical JWH 203 NpentnoIc ACD meta
SDP
Catalog No. 502969331
Encompass_Preferred
An internal standard for the quantification of JWH 203 N-pentanoic acid metabolite by GC- or LC-MS
Catalog No. 50-296-9331 Supplier Cayman Chemical Supplier No. 14369500 UG
An internal standard for the quantification of JWH 203 N-pentanoic acid metabolite by GC- or LC-MS