Learn More
Cayman Chemical JWH 210 2-ethylnaphthyl isomer, Purity: ≥98%, CAS: 1427325-79-8, 10 mg
SDP
Catalog No. 502893030
Encompass_Preferred
A positional isomer of JWH 210, having the ethyl side chain at the 2 position rather than at the 4 position of the naphthyl group; intended for forensic purposes
Catalog No. 50-289-3030 Supplier Cayman Chemical Supplier No. 900103810 MG
A positional isomer of JWH 210, having the ethyl side chain at the 2 position rather than at the 4 position of the naphthyl group; intended for forensic purposes