Learn More
Cayman Chemical JWH 210 3ethylnaphthyl 10mg
SDP
Catalog No. 502893375
Encompass_Preferred
A positional isomer of JWH 210, having the ethyl side chain at the 3 position rather than at the 4 position of the naphthyl group; intended for forensic purposes
Catalog No. 50-289-3375 Supplier Cayman Chemical Supplier No. 900103910 MG
A positional isomer of JWH 210, having the ethyl side chain at the 3 position rather than at the 4 position of the naphthyl group; intended for forensic purposes