Learn More
Cayman Chemical JWH 210 5ethylnaphthyl 10mg
SDP
Catalog No. 502894031
Encompass_Preferred
A positional isomer of JWH 210, having the ethyl side chain at the 5 position rather than at the 4 position of the naphthyl group; intended for forensic purposes
Catalog No. 50-289-4031 Supplier Cayman Chemical Supplier No. 900104010 MG
A positional isomer of JWH 210, having the ethyl side chain at the 5 position rather than at the 4 position of the naphthyl group; intended for forensic purposes