Learn More
Cayman Chemical JWH 210 5hydroxyIndl meta 10mg
SDP
Catalog No. 502893291
Encompass_Preferred
An expected metabolite of JWH 210, detectable both in serum and urine; intended for research and forensic applications
Catalog No. 50-289-3291 Supplier Cayman Chemical Supplier No. 900077110 MG
An expected metabolite of JWH 210, detectable both in serum and urine; intended for research and forensic applications