Learn More
Cayman Chemical JWH 210 7ethylnaphthyl 10mg
SDP
Catalog No. 502895429
Encompass_Preferred
A positional isomer of JWH 210, having the ethyl side chain at the 7 position rather than at the 4 position of the naphthyl group; intended for forensic purposes
Catalog No. 50-289-5429 Supplier Cayman Chemical Supplier No. 900104210 MG
A positional isomer of JWH 210, having the ethyl side chain at the 7 position rather than at the 4 position of the naphthyl group; intended for forensic purposes