Learn More
Cayman Chemical JWH 210d9 GC MS Standards
SDP
Catalog No. 502887994
Encompass_Preferred
An internal standard for the quantification of JWH 210 by GC- or LC-MS
Catalog No. 50-288-7994 Supplier Cayman Chemical Supplier No. 10510500 UG
An internal standard for the quantification of JWH 210 by GC- or LC-MS