Learn More
Cayman Chemical JWH 210d9 GC MS Standards
SDP
Supplier:  Cayman Chemical 105101 MG
Encompass_Preferred
An internal standard for the quantification of JWH 210 by GC- or LC-MS
Catalog No. 50-288-7877