Learn More
Cayman Chemical JWH 250 5-hydroxyindole metabolite, Purity: ≥98%, CAS: 1379604-67-7, 10 mg
SDP
Catalog No. 502893458
Encompass_Preferred
Expected to be a metabolite of JWH 250 that would be detectable both in serum and in urine; intended for research and forensic applications
Catalog No. 50-289-3458 Supplier Cayman Chemical Supplier No. 900076610 MG
Expected to be a metabolite of JWH 250 that would be detectable both in serum and in urine; intended for research and forensic applications