Learn More
Cayman Chemical JWH 250 N-(4-hydroxypentyl) metabolite, Purity: ≥98%, CAS: 1427521-38-7, 10 mg
SDP
Catalog No. 502951375
Encompass_Preferred
Expected to be a cytochrome P450 phase I metabolite of JWH 250, detectable both in serum and urine; intended for research and forensic applications
Catalog No. 50-295-1375 Supplier Cayman Chemical Supplier No. 1093910 MG
Expected to be a cytochrome P450 phase I metabolite of JWH 250, detectable both in serum and urine; intended for research and forensic applications