Learn More
Cayman Chemical JWH 250 N- 5-hydroxypentyl m
SDP
Supplier:  Cayman Chemical 900076710 MG
Encompass_Preferred
Expected metabolite of JWH 250 that would be detectable both in serum and in urine; intended for research and forensic applications
Catalog No. 50-289-3066