Learn More
Cayman Chemical JWH 250 N 5hydroxypentyl m
SDP
Catalog No. 502951669
Encompass_Preferred
An internal standard for the quantification of JWH 250 N-(5-hydroxypentyl) metabolite by GC- or LC-MS
Catalog No. 50-295-1669 Supplier Cayman Chemical Supplier No. 11474100 UG
An internal standard for the quantification of JWH 250 N-(5-hydroxypentyl) metabolite by GC- or LC-MS