Learn More
Cayman Chemical JWH 251 4-methylphenyl isomer, Purity: ≥98%, CAS: 864445-41-0, 10 mg
SDP
Catalog No. 502895720
Encompass_Preferred
Differs from JWH 251 by having the methyl group at the 4 position, rather than the 2 position, on the phenyl group
Catalog No. 50-289-5720 Supplier Cayman Chemical Supplier No. 900102210 MG
Differs from JWH 251 by having the methyl group at the 4 position, rather than the 2 position, on the phenyl group